Learn More
Invitrogen™ NORE1 Monoclonal Antibody (OTI1A11)

Description
NORE1 Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | NORE1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI1A11 |
| Concentration | 0.47 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RASSF5 |
| Gene Accession No. | Q8WWW0 |
| Gene Alias | 1300019G20Rik; AU042887; Maxp1; MGC10823; MGC17344; new ras effector 1; NORE1; Nore1A; NORE1B; novel ras effector 1; protein interacting with guanine nucleotide exchange factor; Rap1-binding protein; RAPL; Ras association (RalGDS/AF-6) domain family 5; Ras association (RalGDS/AF-6) domain family member 5; Ras association domain family member 5; ras association domain-containing protein 5; Ras effector-like protein; RASSF3; RASSF5; regulator for cell adhesion and polarization enriched in lymphoid tissue; regulator for cell adhesion and polarization enriched in lymphoid tissues; RP11-343H5.1; tumor; tumor suppressor RASSF3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.