Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NORE1 Monoclonal Antibody (OTI1H2)
GREENER_CHOICE

Catalog No. PIMA525439
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525439 Supplier Invitrogen™ Supplier No. MA525439
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NORE1 Monoclonal Antibody for Western Blot, Flow

This gene is a member of the Ras association domain family. It functions as a tumor suppressor, and is inactivated in a variety of cancers. The encoded protein localizes to centrosomes and microtubules, and associates with the GTP-activated forms of Ras, Rap1, and several other Ras-like small GTPases. The protein regulates lymphocyte adhesion and suppresses cell growth in response to activated Rap1 or Ras. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NORE1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1H2
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RASSF5
Gene Accession No. Q8WWW0
Gene Alias 1300019G20Rik; AU042887; Maxp1; MGC10823; MGC17344; new ras effector 1; NORE1; Nore1A; NORE1B; novel ras effector 1; protein interacting with guanine nucleotide exchange factor; Rap1-binding protein; RAPL; Ras association (RalGDS/AF-6) domain family 5; Ras association (RalGDS/AF-6) domain family member 5; Ras association domain family member 5; ras association domain-containing protein 5; Ras effector-like protein; RASSF3; RASSF5; regulator for cell adhesion and polarization enriched in lymphoid tissue; regulator for cell adhesion and polarization enriched in lymphoid tissues; RP11-343H5.1; tumor; tumor suppressor RASSF3
Gene Symbols RASSF5
Host Species Mouse
Immunogen Full length human recombinant protein of RASSF5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83593
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.