Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Norepinephrine Transporter (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577494
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Norepinephrine Transporter [NET, or noradrenaline transporter (NAT)] is a monoamine transporter that transports the neurotransmitter noradrenaline from the synapse back to its vesicles for storage until later use. It also appears to transport the neurotransmitter dopamine in the same way, but to a lesser degree. Studies have shown a decrease in NET levels in the locus coeruleus in patients diagnosed with major depression. Cocaine, amphetamines and many therapeutic antidepressants, such as the SNRIs (serotonin-norepinephrine reuptake inhibitors) and the tricyclic antidepressants (TCAs) act to raise noradrenaline. Furthermore, deficits in the NET gene have been associated with ADHD.
Specifications
Norepinephrine Transporter (extracellular) | |
Polyclonal | |
Unconjugated | |
Slc6a2 | |
Norepinephrine transporter, SLC6A2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20538, 83511 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O55192 | |
Slc6a2 | |
Peptide (C)KLLNASVLGDHTKYSK, corresponding to amino acid residues 189-204 of mouse Noradrenaline Transporter | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction