Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Norepinephrine Transporter Mouse anti-Mouse, Rat Monolconal, Abnova™

Catalog No. 89035525 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-525 Supplier Abnova Corporation Supplier No. MAB14127

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against synthetic peptide of mouse Norepinephrine Transporter.

  • High specificity
  • High reactivity

Specifications

Antigen Norepinephrine Transporter
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of mouse Norepinephrine Transporter.
Dilution Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In 0.01 M HEPES, 0.15 M NaCl, pH 7.5 (50% glycerol, 100 ug/mL BSA).
Gene Accession No. O55192
Host Species Mouse
Immunogen A synthetic peptide corresponding to N-terminus of mouse Norepinephrine Transporter.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.