Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Norovirus capsid G1 Monoclonal Antibody (S57S)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518237
Description
This antibody does not cross-react with capsid G2. Purity is >90%. Recommended pairs for ELISA: MA5-18237 (capture) and MA5-18239 (conjugated, detection). This antibody can also be used as a detection antibody in lateral flow applications.

Specifications
Norovirus capsid G1 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Lateral Flow, Western Blot | |
S57S | |
Unconjugated | |
Norwalk virus | |
Norovirus recombinant protein. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction