Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Norovirus GII.17 VLPS Monoclonal Antibody (4937)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA547539

Catalog No. PIMA547539


Only null left
Add to Cart

Description

Description

Specificity: Reactive with GII.17 VLPS. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

Norovirus is a positive strand RNA virus with three ORFs. These open reading frames encode a large polyprotein that produces two structural proteins, VP1 and VP2, and non-structural proteins through proteolytic cleavage.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Norovirus GII.17 VLPS
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Virus
Antibody
IgG1
ELISA
4937
Unconjugated
GII.17; VLPS
Proprietary sequence.
100 μg
Primary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.