Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Norovirus (MNV-1) Mouse anti-Virus, Unconjugated, Clone: 5C4.10, MilliporeSigma™

Catalog No. MABF2097100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2097-100 100 μg
MABF2097-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2097-100 Supplier MilliporeSigma Supplier No. MABF2097100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Norovirus (MNV-1), clone 5C4.10, Cat. No. MABF2097, is a mouse monoclonal antibody that detects murine Norovirus capsid protein and has been tested for use in ELISA and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Norovirus (MNV-1)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5C4.10
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias MNV-1;Murine Norovirus
Host Species Mouse
Immunogen Infectious and UV-inactivated murine norovirus-1.
Molecular Weight of Antigen ∽57 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone 5C4.10 specifically recognizes the S domain portion of capsid protein of murine Norovisrus.
Target Species Virus
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.