Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Norovirus VP1 Recombinant Rabbit Monoclonal Antibody (HL2177)
GREENER_CHOICE

Catalog No. FEMA550420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550420 Supplier Invitrogen™ Supplier No. MA550420
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Norovirus VP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Norovirusis a very contagious virus that causes vomiting and diarrhea. People of all ages can get infected and sick withnorovirus.Norovirusspreads easily! People withnorovirusillness can shed billions ofnorovirusparticles. Mutations in the protruding domain 2 (P2 domain) of the norovirus major capsid protein (VP1) result in the emergence of various NOV variants, however, it is unclear whether the minor capsid protein (VP2) also affects the generation of VP1 variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Norovirus VP1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2177
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Norovirus
Host Species Rabbit
Immunogen Synthetic peptide of Norovirus VP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.