Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Norovirus VP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117420
Description
Centrifuge briefly prior to opening.
Norovirusis a very contagious virus that causes vomiting and diarrhea. People of all ages can get infected and sick withnorovirus.Norovirusspreads easily! People withnorovirusillness can shed billions ofnorovirusparticles. Mutations in the protruding domain 2 (P2 domain) of the norovirus major capsid protein (VP1) result in the emergence of various NOV variants, however, it is unclear whether the minor capsid protein (VP2) also affects the generation of VP1 variants.
Specifications
| Norovirus VP2 | |
| Polyclonal | |
| Unconjugated | |
| Norovirus | |
| Recombinant protein encompassing a sequence within the center region of Norovirus VP2 ( GII.17) (Norovirus (Hu/GII/CN/2014/GII.17/1401169)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.57 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction