Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Norrin/NDP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | Norrin/NDP |
---|---|
Dilution | Western Blot Reactivity reported in (PMID: 29860944)., Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Norrin/NDP Polyclonal specifically detects Norrin/NDP in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Norrin/NDP | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
EVR2, exudative vitreoretinopathy 2 (X-linked), FEVR, ND, Norrie disease (pseudoglioma), Norrie disease protein, norrin, X-linked exudative vitreoretinopathy 2 protein | |
NDP | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot Reactivity reported in (PMID: 29860944)., Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Neuroscience, Vision | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
4693 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TDSSFIMDSDPRRCMRHHYVDSISHPLYKCSSKMVLLARCEGHCSQASRSEPLVSFSTVLKQPFRSSCHCCRPQTSKLKALRLRCSGGMRLTATYRYILSCHCEECNS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title