Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nortestosterone Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175005
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175005 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175005 Supplier Invitrogen™ Supplier No. PA175005
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 0.25 μg/mL. Sensitivity: 10 ng/mL 17alpha-19 Nortestosterone produces 93% inhibition in a competitive ELISA employing 17α-Nortestosterone polyclonal antibody.

Nandrolone is a synthetic, anabolic steroid analog of testosterone. Similar to testosterone, nortestosterone (19-Nortestosterone) binds to and activates specific nuclear receptors. This agent may be used for testosterone replacement therapy in hypogonadal men, in HIV-wasting syndrome, and in other conditions in order to increase nitrogen retention and fat-free muscle mass. [National Center for Biotechnology Information. PubChem Compound Database; CID=9904].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nortestosterone
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias 19-Norandrostenolone; 19-Nortestosterone; C18H26O2; Nandrolone; Norandrostenolone; Nortestosterone
Host Species Sheep
Immunogen 17a-19-Nortestosterone(3)-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.