Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ 5500 Series Convenience Pack Respirators

Catalog No. 19999225
Click to view available options
Cartridge Type:
Defender™ Multi-Contaminant Cartridges
Organic Vapor
Organic Vapor, Acid Gas
P100 Cartridge
Size:
Small
Medium
Large
Includes:
Multi-Contaminant Cartridges and P100 Filters
OV Cartridge
OV Cartridge, N95 Filter
OV Cartridge, P100 Particulate Filters
OV Cartridge, R95 Filter
OV and AG Cartridges
OV and AG Cartridges, P100 Particulate Filters
P100 Particulate Filters

Elastomeric half mask respirators offer a wide contoured sealing with a secure fit. Choose from several filter options depending on specific application.

  • Low-maintenance, reusable, economical
  • Soft, comfortable elastomer facepiece
  • For use with North cartridges, filters and accessories
  • Incorporates the anatomic design of the North 7700 Series respirators
  • Requires very little maintenance
  • Contoured sealing flange and cradle suspension system combine to ensure proper positioning
  • All parts are replaceable, extending service life
  • Easy-to-order 5500 Series Convenience Packs include most commonly used cartridges and filters
  • Available in a range of sizes and cartridge/filter combinations
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Organic Vapor
Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Filter Type P100
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Medium
Facepiece Material Thermoplastic Elastomer
For Use With (Application) Agriculture, food services, government, municipal service
Includes OV Cartridge, P100 Particulate Filters
Model 5581P100M
Weight (English) 0.9 lb.
Weight (Metric) 0.4 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.