Learn More
Honeywell™ 5500 Series Half-Mask Air Purifying Respirators
Shop All Protective Industrial Products Inc. ProductsDescription
- Easy-to-clean facepiece conforms gently to wearer's face and is made from a soft elastomer that resists distortion
- 'One-piece' cradle suspension system is easily removed for cleaning and provides a comfortable, secure fit with no slipping
- Contoured sealing flange eliminates pressure points
- Extended side flanges ensure secure seal even when wearer is talking
- Exhalation valve assembly provides low breathing resistance and permits easy, positive pressure fit check without removing cover
- Headband straps are adjustable and virtually impossible to lose or misassemble; headband yoke improves peripheral vision by allowing cartridges to be lower and farther back
- Direct cartridge-to-facepiece seal eliminates risk of improper seal and reduced protection due to lost or worn sealing gaskets
- Requires limited maintenance and available in three sizes that accept North cartridges and filters
Compliance
NIOSH approved under 42 CFR Part 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters.
Specifications
Specifications
| Certifications/Compliance | MSHA |
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Certifications (OSHA) | OSHA 29 CFR Part 1920.134 |
| Connection Type | Threaded |
| Color | Gray |
| Material | Thermoplastic Elastomer |
| Product Type | Half-Mask Respirator |
| Size | Medium |
| For Use With (Equipment) | North™ Cartridge, Filter and Accessories |
| Model | 550030M |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.