Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ North™ 7600 Series Silicone Full Facepiece Air-Purifying Respirator
Click to view available options
Size:
Medium/Large
Small
Includes:
Welding Attachment
Without Welding Attachment
Description
Requires
Compatible with North cartridges, filters and accessories.
- Protects eyes and face against irritating gases, vapors and flying particles
- Available with or without welding attachment
- Dual-flange silicone seal conforms to facial features
- Speaking diaphragm clarifies conversations enhancing safety and other communication
- Hard-coated polycarbonate lens provides over 200° field of vision
- Oral nasal cup reduces fogging and lessens dead air space, improving worker comfort
- Five-strap harness provides a comfortable facepiece-to-face seal
- Chin cup helps proper positioning of facepiece
- Neck strap allows wearer to park respirator when not in use
- Threaded cartridge connector secures equipment to avoid accidental disconnections
- Can be adapted to PAPR or CF-SAR; compatible with PA 700 Series PAPR and CF2000 Series SAR
Certifications
NIOSH approved under 42 CFR 84 when used with appropriate cartridges and filters.
Compliance
NIOSH approved under 42 CFR 84 for use w/ a variety of respiratory hazards when used as designed with approp N-Series cartridges and filters in conjunc with OSHA fit testing reqts. Meets OSHA 29 CFR Part 1920.134; lens meets ANSI Z87.1 reqts.
Respirator, Full, 7600, Silicone, APR
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Connection Type | Threaded |
| Color | Black |
| Material | Silicone |
| Product Type | Full Facepiece Respirator |
| Size | Small |
| Facepiece Material | Silicone |
| Includes | Welding Attachment |
| Latex Free | Yes |
| Material (Lens) | Polycarbonate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction