Learn More
Honeywell™ North™ A800 Series Safety Glasses
Catalog No. 191600226 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Clear
Gray
Lens Tint:
Clear
Silver Mirror, Indoor/Outdoor
TSR Gray
Lens Coating:
Anti-fog
Scratch-resistant
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-160-0226 Clear Clear Anti-fog
19-160-0224 Gray TSR Gray Scratch-resistant
19-160-0225 Gray Silver Mirror, Indoor/Outdoor -
19-160-0223 Clear Clear Scratch-resistant
19-160-0227 Gray TSR Gray Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-160-0226 Supplier Honeywell™ Supplier No. A805
Add to Cart
Add to Cart
This item is not returnable. View return policy
Aggressive sport styling; soft temple inserts with extra flex for comfort
  • 9-base polycarbonate hardcoated lenses provide 180° field of distortion-free vision; block 99.9% UV
  • Soft nosepiece conforms to various nose shapes for nonslip fit
  • Available with scratch-resistant hardcoat or Fog-Ban™ antifog coating
Compliance
Meet or exceed ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Anti-fog
Certifications/Compliance Meets or exceeds ANSI Z87.1-2003 standards.
Lens Tint Clear
Frame Color Clear
Lens Material Polycarbonate