Learn More
Honeywell™ North™ A900 Series Reader Magnifiers
Catalog No. 191601207
Click to view available options
Lens Tint:
Clear
Magnification Power:
+2.50
This item is not returnable. View return policy
This item is not returnable. View return policy
For close-up inspection, fine detail work and reading small print
  • Wraparound eight-base optically perfected lenses provide panoramic 180-degree distortion-free vision
  • Soft padded tips for less temple fatigue
  • Soft, nonslip nose bridge for reduced slippage
  • UV protection: up to 99.9%
  • Black frames, hardcoat lenses
  • Free FlexiCord™ keeps eyewear handy and suits workers who need to switch routinely between regular and magnifier eyewear
Compliance
Meets ANSI Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Hardcoat
Certifications/Compliance ANSI Z87.1-2010
Lens Tint Clear
Product Type Reader Magnifiers
For Use With (Application) For close-up inspection, fine detail work and reading small print
Frame Color Black
Magnification Power +2.50
UV Protection 0.999
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.