Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ EXC Cap Mounted Ear Muff

Catalog No. 19130343 Shop All MSA Products
Change view
Click to view available options
Hard Hat-Attached Ear Muffs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-130-343 Hard Hat-Attached Ear Muffs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-130-343 Supplier MSA™ Supplier No. 10061230

EXC Cap Mounted Ear Muffs are designed to be used in conjunction with MSA™ Slotted Caps and are comfortable for all-day wear.

  • Hard Hat-Attached style is designed to be worn with MSA slotted hard hats
  • Includes brackets for mounting
  • Comfortable for all-day wear
  • Unique injection molded inserts provides maximum space for ears
  • Features a spring design that offers low pressure tension
  • Gray

Specifications

Color Black, Gray
ANSI S3.1974 Yes
CSA Z94.2 Class A
Cushion Color Black
For Use With (Equipment) MSA™ Slotted Hard Hats
For Use With (Application) General industry, manufacturing, repair and maintenance, construction, oil and gas, forestry, shipbuilding, mining
High Visibility No
Model 10061230
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type SoundControl
Dielectric No
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.