Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ First Aid Kit and Replacement Items

Catalog No. p-119660
Click to view available options
Product Type:
Nitrile Gloves

Specifications

For Use With (Application) First Aid
Length (English) 9.5 in.
Length (Metric) 24 cm
Product Type Nitrile Gloves
Color Blue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.