Learn More
Honeywell™ North™Flexible Low Profile P100 Filters
Catalog No. 19003263 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Product Type:
Filter
Respirator Filter Plus Odor Relief
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-003-263 Filter
19-161-841 Respirator Filter Plus Odor Relief
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-003-263 Supplier Honeywell™ Supplier No. 75FFP100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thin, high-performance respirator filters are P100 low profile, flexible filters. Available with or without nuisance odor relief.
Cartridges for use with 5500, RU8500 and 7700 Series half masks as well as 5400, RU6500 and 7600 Series full facepieces.
Specifications
Cartridge Type Filter Cartridge
Certifications/Compliance NIOSH
Color Magenta
Product Type Filter
Quantity 50/Cs.