Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Grip N™ Aramid Fiber Cut-Resistant Gloves

Catalog No. p-141423
Click to view available options
Size:
Universal
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless knit aramid fiber cut-resistant gloves feature a nitrile 'N' coating on both sides. Complies with USDA and FDA regulations, 21 CFR for food processing.

  • Nitrile “N” coating improves grip and extends life due to better abrasion resistance
  • Knit construction provides comfort and minimal hand fatigue
  • Machine washable or dry cleanable
  • Ambidextrous design gives wearer four sides instead of two to further enhance value
  • Blended design
  • Aramid fibers positioned on outside for cut/slash protection and cotton/polyester on inside for worker comfort

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA 21 CFR, OSHA 29 CFR 1915.157
Construction Seamless Knit
Color Yellow, Rust
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Universal
EN388 Standard EN 388
For Use With (Application) Fabricating, manufacturing, sharp part handling, sheet metal handling
Gender Men's
Glove Cuff Style Knit
Model 52/6647M
Series Grip N™
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.