Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ North™ HM500 Series Half Mask
Click to view available options
Size:
Medium
Description
- Lighter than other similar masks with cartridges available on the market
- Softer material for better touch and seal with skin, for long-time use
- Professional design for universal fit
- Advanced over-molding manufacturing and design made for intuitive assembly and disassembly
- Drop-down function replaces need to doff hardhat or earmuff for quick work break (Fisher cat #'s 19-185-597, 598 and 599)
- Easy cleaning and maintenance
Specifications
Specifications
| Connection Type | Threaded |
| Color | Black |
| Material | Thermoplastic Elastomer |
| Product Type | Half-Mask Respirator |
| Size | Medium |
| With Exhalation Valve | Yes |
| For Use With (Application) | Oil and gas, pharmaceutical, general manufacturing, metal industry, construction |
| For Use With (Equipment) | N-Series filters and cartridges |
| Latex Free | Yes |
| Model | HM501TM |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction