Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ HM500 Series Half Mask

Catalog No. 19185596
Click to view available options
Size:
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Threaded half-mask is lightweight, durable and soft with easy operation features. Filters and cartridges not included.

  • Lighter than other similar masks with cartridges available on the market
  • Softer material for better touch and seal with skin, for long-time use
  • Professional design for universal fit
  • Advanced over-molding manufacturing and design made for intuitive assembly and disassembly
  • Drop-down function replaces need to doff hardhat or earmuff for quick work break (Fisher cat #'s 19-185-597, 598 and 599)
  • Easy cleaning and maintenance
TRUSTED_SUSTAINABILITY

Specifications

Connection Type Threaded
Color Black
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Large
With Exhalation Valve Yes
For Use With (Application) Oil and gas, pharmaceutical, general manufacturing, metal industry, construction
For Use With (Equipment) N-Series filters and cartridges
Latex Free Yes
Model HM501TL
Strap Attachment Type Non drop-down
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.