Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Light Task Plus 3™ Dyneema™ Cut-Resistant Gloves, Polyurethane Coated

Catalog No. 19130271C
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Medium-weight cut-resistant glove that offers a great compromise between mechanical protection and wearer comfort.

Suited for material handling and assembly work involving sharp objects like sheet metal, metal shavings, glass and metal parts—anywhere mechanical hazards exist

  • Seamless Dyneema liner is up to 15 times stronger than steel (weight for weight) and outperforms aramid fibers in cut and abrasion resistance
  • Dyneema shell is stretchy and form fitting, allowing dexterity
  • PU coating provides abrasion resistance, but is light enough to ensure workers retain dexterity and tactility
  • Gray color hides dirt, making it a good choice for many tough and dirty applications
  • Uncoated back of glove assures ventilation and comfort
  • Contains no reaction causing, natural rubber proteins
  • Cuff over-stitching is color-coded by size to aid sorting after laundering

TRUSTED_SUSTAINABILITY

Specifications

Construction Seamless Knit
Color Gray, White
Cut Resistance Rating ANSI A1
Material Dyneema
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388
For Use With (Application) sheet Metal fabrication, machine shops, Glass industry, automotive and other assembly work
Glove Cuff Style Knit
Model NFD16G/9L
Series NorthFlex Light Task Plus 3™
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.