Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Low-Profile Cartridges and Cartridge/P100 Filter Combinations

Catalog No. 19003263F Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Cartridge Type:
Acid Gas Cartridge
Ammonia, Methylamine Cartridge
Cartridge/Filter Assembly
Filter Cartridge
Low Profile Particulate Cartridge
Mercury Vapor and Chlorine Cartridge
Organic Vapor Cartridge
Organic Vapor, Acid Gas Cartridge
Color:
Black
Green/Magenta
Magenta/White
Magenta/Yellow
Olive
Olive/Magenta
White
Yellow
Quantity:
12/Cs.
18/Cs.
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Color Quantity
19-003-263F Acid Gas Cartridge Magenta/White 12/Cs.
19-003-263H Filter Cartridge Magenta/Yellow 12/Cs.
19-003-263J Ammonia, Methylamine Cartridge Green/Magenta 12/Cs.
19-003-299B Acid Gas Cartridge White 18/Cs.
19-003-300B Organic Vapor, Acid Gas Cartridge Yellow 18/Cs.
19-003-298B Organic Vapor Cartridge Black 18/Cs.
19-003-309B Mercury Vapor and Chlorine Cartridge Olive 18/Cs.
19-003-310B Cartridge/Filter Assembly Olive/Magenta 12/Cs.
19-003-312B Low Profile Particulate Cartridge Olive 18/Cs.
17-622-14 Organic Vapor Cartridge Black 12/Cs.
19-003-313B Filter Cartridge Olive/Magenta 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19-003-263F Supplier Honeywell™ Supplier No. 7582P100L
Add to Cart
Add to Cart

Satisfy numerous industrial applications

  • All North air-purifying cartridges are approved with all North air-purifying half masks and full-facepiece respirators
  • Air-purifying respirators with cartridges provide respiratory protection against gases and vapors by using sorbents to purify the inhaled air
  • Cartridges without filters do not remove dust, fibers, and other particles

Certifications

All North cartridges, filters, and cartridge/filter combinations are NIOSH certified to 42CFR84 standards when used with North respirator facepieces.

Compliance

NIOSH approved.

Specifications

Cartridge Type Acid Gas Cartridge
Certifications/Compliance NIOSH, 42CFR84
Color Magenta/White
Product Type Cartridge
For Use With (Application) Industrial applications
For Use With (Equipment) Respirator
Quantity 12/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.