Learn More
Honeywell™ North™ Low-Profile Cartridges and Cartridge/P100 Filter Combinations
Shop All Protective Industrial Products Inc. ProductsDescription
- All North air-purifying cartridges are approved with all North air-purifying half masks and full-facepiece respirators
- Air-purifying respirators with cartridges provide respiratory protection against gases and vapors by using sorbents to purify the inhaled air
- Cartridges without filters do not remove dust, fibers, and other particles
Certifications
All North cartridges, filters, and cartridge/filter combinations are NIOSH certified to 42CFR84 standards when used with North respirator facepieces.
Compliance
NIOSH approved.
Specifications
Specifications
| Cartridge Type | Mercury Vapor and Chlorine Cartridge |
| Certifications/Compliance | NIOSH, 42CFR84 |
| Color | Olive |
| Product Type | Cartridge |
| For Use With (Application) | Industrial applications |
| For Use With (Equipment) | Respirator |
| Quantity | 18/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.