Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Nitri Task™ Nitrile Coated Nylon Gloves

Catalog No. p-3349626
Click to view available options
Size:
Small
Medium
Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight, 13-gauge gloves feature nitrile coating on the palm and fingertips for enhanced grip capabilities.

  • Lightweight, 13-gauge nylon
  • Nitrile coating on the palm and fingertips
  • Knit wrist prevents dirt and debris from entering the glove
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color Gray, White
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Automotive Assembly, General Maintenance, Manufacturing, and Small Parts Handling
Glove Cuff Style Knit
Series NF13
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.