Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ NorthFlex Cold Grip™ PVC-Coated Polyamide Insulated Gloves

Catalog No. 17805010 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-805-010 Large
19-045-561 Medium
17-805-011 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-805-010 Supplier Honeywell™ North™ Supplier No. NF11HD/9L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cold Grip™ Gloves provide workers with comfort and enhanced grip capabilities, while also offering protection from temperatures as low as -26°C (-15°F).

  • Heavyweight insulated gloves without sacrificing comfort and dexterity
  • Polyamide yarn reduces linting for the sensitive applications
  • Black PVC coating covers the palm and fingers for enhanced grip capabilities
  • Hydropellent Technology repels liquids to provide a dry, firm grip
  • Ideal for handling damp, oily, or greasy parts in cold conditions
  • Recommended for temperatures down to -26°C (-15°F)
  • Launderable

Specifications

Coating Material PVC
Color Black
Exterior Finish Fully Textured
Hand Specific
Material Polyamide
Size Large
For Use With (Application) Construction, Logistics, and Maintenance
Glove Cuff Color Blue
Glove Cuff Style Knit
Liner Material Synthetic Insulation
Series NF11HD
Temperature (English) Min -15°F
Temperature (Metric) Min. -26°C
Yarn Gauge 7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.