Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Light Task Plus II™ Polyurethane Coated Gloves with Dyneema™ Fibers

Catalog No. 191501004
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight, 13-gauge Dyneema™ fiber gloves provide protection against cuts and slashes. Polyurethane coating ensures high tactile sensitivity and abrasion resistance.

  • Seamless Dyneema™ cut-resistant fibers
  • Polyurethane coating on the palm and fingers
  • Knit wrist helps prevent dirt and debris from entering the glove
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A2
Hand Specific
Material Dyneema
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Sheet Metal Handling, and Metal Fabrication
Glove Cuff Style Knit
Series NFD15
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.