Learn More
Honeywell™ Sperian™ One-Fit™ N95 Particulate Respirators
Universal sized disposable respirators fit most face shapes and sizes.
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-4373
Color White
Design Type Cup/Molded
Disposable Single-use
View More Specs
Products 1
Catalog Number Mfr. No. Size Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Size Quantity Price Quantity & Availability  
19-034-920
View Documents
Honeywell™
14110444
Standard 1 Ea. N/A N/A N/A
Description
Description
  • Electrostatic nonwoven polypropylene and polyester materials; lightweight construction allows for longer wear time and less fatigue
  • Durable filtration media ensures filtration efficiency of 95% against non-oil airborne particles
  • Protection against airborne pathogens and M. tuberculosis as well as certain airborne particulates and pandemic diseases such as H5N1 Avian Flu and SARs
W1400 Flat Fold Mask
  • Bonded wire nose clip with open cell foam nose seal
  • Low-profile design offers compatibility with spectacles
NBW95 and NBW95V Form Fitted Masks
  • Strong and rigid outer shell prevents mask from collapse in high humidity or moist environments
  • Molded nose bridge fits comfortably and is compatible with spectacles
Specifications
Specifications
N - Not Resistant to Oil
White
Single-use
Polypropylene, Polyester
Yes
Yes
Dual Welded Head Straps
NIOSH 42 CFR 84, N95, approval number: TC-84A-4373
Cup/Molded
N95
General Purpose Particulate Respirator
95%
Elastic
No
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.