Learn More
Honeywell™ North™ Panther Hip-Pac Respirator
This product provides air from a remote air source for entry into and escape from IDLH atmospheres. The escape cylinder is available in 5-, 10- and 15-minute durations.
Supplier: Honeywell™ P968477
Description
The Pressure Demand SAR with Escape Cylinder (Hip-Pac) offers the same reliable facepieces and second stage regulators as the Cougar and 1997-style Panther SCBAs. It allows the user to comfortably wear the respirator to enter an IDLH environment while breathing from a remote air source. Should the air supply be interrupted, the user can switch to the cylinder for escape.
Specifications
NIOSH | |
10 min. | |
Chemical, construction, food services, manufacturing, military, mining, oil and gas, pharmaceutical |
Standard Escape Respirator | |
10 min. | |
3000 psi aluminum cylinder, silicone twenty plus facepiece, regulator, medium, black, medium nose cup, silicone headstrap |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.