Learn More
Honeywell™ North™ High Performance Face Shield Headgear - 4 in. Crown
Catalog No. 189994459
Change view
Click to view available options
:
Heat resistant browguard; w/Ratchet adjustment and sweatband
Catalog No.
18-999-4459 Heat resistant browguard; w/Ratchet adjustment and sweatband
1 options
This item is not returnable. View return policy
Catalog No. 18-999-4459
Supplier: Honeywell™ 11380048
This item is not returnable. View return policy
Adjustable ratchet headgear fits a variety of hat sizes
  • Sturdy browguard is constructed of heat-resistant thermoplastic polymer
  • Brim overhang keeps fluids from splashing or dripping inside window
  • Headgear fits hat sizes from 61/2 to 8
Basic eye protection must be worn under face shields to protect from flying particles and hazardous splashes.
Order Info
Windows sold separately (see below).
Compliance
Meets ANSI Z87.1+ and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Fits a variety of hat sizes
Includes Ratchet adjustment and sweatband