Learn More
Honeywell™ North™ Replacement Cylinders for SCBA Units
Specifications
Specifications
| Product Type | Air Cylinder |
| Duration | 30 min. |
| Pressure | 2216 psi |
| For Use With (Application) | Chemical resistance and allows protective eyewear to be worn |
| For Use With (Equipment) | Scba |
| Includes | A nylon harness and a compact side mounted demand valve |
| Material | Aluminum |