Learn More
Honeywell™ North™ Respirator Cartridges and Filters
Catalog No. 19003301A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
For Use With (Equipment):
All North™ air-purifying facepiece respirators
North™ 5400, 5500, 7600 and 7700 respirators
North™ full or half mask respirators
Product Type:
Cartridge
N95 Non-Oil Particulate Filter
N99 Non-Oil Particulate Filter
Particulate Filter
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
19-003-301A North™ full or half mask respirators Cartridge
17-622-7 North™ 5400, 5500, 7600 and 7700 respirators N95 Non-Oil Particulate Filter
19-003-311B North™ full or half mask respirators Particulate Filter
19-003-305 All North™ air-purifying facepiece respirators N99 Non-Oil Particulate Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-003-301A Supplier Honeywell™ Supplier No. N75004L
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cartridges for use with 5500, RU8500 and 7700 Series half masks as well as 5400, RU6500 and 7600 Series full facepieces.
  • Filter assemblies for use with facepiece alone or with gas and vapor cartridges available
Specifications
Cartridge Type Ammonia, Methylamine Cartridge
Certifications/Compliance NIOSH
Product Type Cartridge
For Use With (Equipment) North™ full or half mask respirators
Quantity 1 Pair