Learn More
Honeywell™ North™ Respirator Cartridges and Filters
Shop All Protective Industrial Products Inc. ProductsDescription
- Filter assemblies for use with facepiece alone or with gas and vapor cartridges available
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Color | Magenta |
| Product Type | Particulate Filter |
| For Use With (Equipment) | North™ full or half mask respirators |
| Quantity | 1 Pair |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.