Learn More
Honeywell™ North™ Unisorb Cartridges
Eliminate the need to stock contaminant-specific cartridges to simplify training, ordering, and inventory. Survivair T-Series Twist-Lock Cartridges provide an audible click indicating a positive lock on connector. NIOSH approved for up to 16 contaminants.
Specifications
Certifications/Compliance NIOSH
View More Specs
Products 2
Catalog Number Mfr. No. Filter Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Filter Type Quantity Price Quantity & Availability  
18-999-1306
View Documents
Honeywell™
T100800
Unisorb Multi-Contaminant Cartridge 6/Pk. N/A N/A N/A
19-003-438
View Documents
Honeywell™
T106010
N95 24/Pk. N/A N/A N/A
Description
Description
These economical cartridges offer lower breathing resistance than many similar products and feature a low profile for better visibility and weight distribution. An audible “locking” bayonet connection confirms cartridge attachment, preventing cross-threading and under/over tightening concerns. And for maximum flexibility, both standard and HEPA cartridges can be used on any Willson ValuAir™ Plus, Premier™ 6800, 6100 half-mask, or 6400/6500 Series full-facepiece respirators.
Used alone, the standard Unisorb cartridge offers protection against a variety of contaminants, including hydrogen fluoride. When paired with a Willson N95 or P95 prefilter and filter retainer (P95 filter sold separately), the Unisorb is approved for use in environments containing dusts, mists, paint spray and pesticides. The Unisorb cartridge with P100 filter does not require a prefilter and also offers NIOSH-approved protection against asbestos-containing dusts and mists.
  1. Standard Unisorb cartridge requires a Willson N95 or P95 prefilter.
  2. Unisorb cartridge with P100 filter only.
  3. Dusts, mists, fumes have a TWA >0.05mg/m.
Specifications
Specifications
NIOSH
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.