Learn More
Honeywell™ North™ Vapor Safety Glasses
Capture sports styling in a painted, metallic blue frame
Specifications
Certifications/Compliance Meet ANSI Z87+ and CSA Z94.3 safety standards
Includes Break-Away Flexicord
Temple Style Sport
Lens Material Polycarbonate
View More Specs
Products 3
Catalog Number Mfr. No. Lens Tint Lens Coating Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Lens Coating Price Quantity & Availability  
19-130-0845
View Documents
Honeywell™
11150901
TSR Gray Hardcoat N/A N/A N/A
19-130-0844
View Documents
Honeywell™
11150900
Clear Hardcoat N/A N/A N/A
19-130-0847
View Documents
Honeywell™
11150905
Clear Anti-fog N/A N/A N/A
Description
Description
Capture sports styling in a painted, metallic blue frame
  • 9–base wrap-around dual lenses and sport temples offer a comfortable secure fit
  • Soft nose bridge prevents slipping
  • Durable polycarbonate lens feature scratch-resistant or Fog-Ban™ antifog coating
  • Adjustable breakaway Flexicord™ included
Compliance
Meet ANSI Z87+ and CSA Z94.3 safety standards
Specifications
Specifications
Meet ANSI Z87+ and CSA Z94.3 safety standards
Break-Away Flexicord
Sport
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.