Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ NorthFlex Red™ Foam PVC Coated Nylon Gloves

Catalog No. 19166665 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-166-665 Small
19-166-666 Medium
19-166-667 Large
19-166-668 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-166-665 Supplier Honeywell™ North™ Supplier No. NF11/7S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight, 15 gauge nylon gloves feature a foam PVC coating on the palm and fingers for enhanced grip capabilities, with an open back for increased ventilation.

  • Foam PVC coating on the palm for enhanced grip capabilities
  • Seamless nylon shell provides a snug fit for comfort and safety
  • Ventilated, uncoated glove back keeps hands cool

Specifications

ANSI Abrasion Rating ANSI 4
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material PVC
Color Red
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Agriculture, Construction, Manufacturing, and Municipal Services
Glove Cuff Style Knit
Series NF11
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.