Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOS1AP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563147
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563147 Supplier Invitrogen™ Supplier No. PA563147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKKSQAMRIV RTVGQAFEVC HKLSLQHTQQ NADGQEDGES ERNSNSSGDP GRQLTGAERA STATAEETDI DAVEVPLPGN DV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

This gene encodes a cytosolic protein that binds to the signaling molecule, neuronal nitric oxide synthase (nNOS). This protein has a C-terminal PDZ-binding domain that mediates interactions with nNOS and an N-terminal phosphotyrosine binding (PTB) domain that binds to the small monomeric G protein, Dexras1. Studies of the related mouse and rat proteins have shown that this protein functions as an adapter protein linking nNOS to specific targets, such as Dexras1 and the synapsins. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOS1AP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOS1AP
Gene Accession No. O75052
Gene Alias 6330408P19Rik; Capon; carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein; C-terminal PDZ domain ligand of neuronal nitric oxide synthase; C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON); C-terminal PDZ ligand of neuronal nitric oxide synthase protein; Kiaa0464; ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain; MGC138500; nitric oxide synthase 1 (neuronal) adaptor protein; nitric oxide synthase 1 adaptor protein; NOS1AP
Gene Symbols NOS1AP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOS1AP. Recombinant protein control fragment (Product #RP-103220).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9722
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.