Learn More
Invitrogen™ NOSIP Polyclonal Antibody

Description
Immunogen sequence: VCAVTRDSLS NATPCAVLRP SGAVVTLECV EKLIRKDMVD PVTGDKLTDR DIIVLQRGGT GFAGSGVKLQ AEKSRPVM Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 95%.
Specifications
Specifications
| Antigen | NOSIP |
| Applications | Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NOSIP |
| Gene Accession No. | Q9Y314 |
| Gene Alias | 2310061K06Rik; CGI-25; E3 ubiquitin-protein ligase NOSIP; eNOS-interacting protein; nitric oxide synthase interacting protein; nitric oxide synthase-interacting protein; Nosip; RING-type E3 ubiquitin transferase NOSIP |
| Gene Symbols | NOSIP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.