Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOSTRIN Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281749AF647
This item is not returnable.
View return policy
Description
NOSTRIN Polyclonal antibody specifically detects NOSTRIN in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
NOSTRIN | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
BM247 homolog, DaIP2, endothelial nitric oxide synthase traffic inducer, ENOS traffic inducer, eNOS-trafficking inducer, Nitric oxide synthase traffic inducer, nitric oxide synthase traffickerMGC20702, nitric oxide synthase trafficking, nostrin, ortholog of mouse disabled 2 interacting proein 2 | |
NOSTRIN antibody was raised against a 17 amino acid peptide near the carboxy terminus of human NOSTRIN. The immunogen is located within amino acids 430 - 480 of NOSTRIN. Amino Acid Squence: SFQARQDDELNLEKGDI | |
0.1 mL | |
Neuroscience, Signal Transduction | |
115677 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction