Learn More
Invitrogen™ NOTCH1 Chimeric Recombinant Mouse Monoclonal Antibody (E6)
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547817
Description
Specificity: An engineered ScFv fragment antibody that binds to both mouse mNRR1 and human hNRR1 but not NRR2. Preparation: This antibody was selected against a fusion protein consisting of EGF domains 1-12 of murine Notch1 fused to a human Fc domain and binds to to the NRR of mouse Notch1.
The notch gene belongs to a family of epidermal growth factor (EGF) like homeotic genes, which encode transmembrane proteins with a variable number of cysteine-rich EGF-like repeats in the extracellular region. Four notch genes have been described in mammals: Notch1, Notch2, Notch3 and Notch4(Int-3), which have been implicated in the differentiation of the nervous system and other structures. The EGF-like proteins Delta and Serrate have been identified as ligands of Notch1. Mature Notch proteins are heterodimeric receptors derived from the cleavage of Notch pre-proteins into an extracellular subunit (NEC) containing multiple EGF-like repeats and a transmembrane subunit including intracellular region (Ntm).
Specifications
NOTCH1 Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P46531, Q01705 | |
Notch1 | |
This antibody was selected against a fusion protein consisting of EGF domains 1-12 of murine Notch1 fused to a human Fc domain and binds to the NRR of mouse Notch1. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 λ |
ELISA, Immunohistochemistry | |
E6 | |
Unconjugated | |
Notch1 | |
9930111A19Rik; AOS5; AOVD1; Drosophila Notch homolog 1 (controlling the the ectodermal and neural cell fate in Drosophila); hN1; lin-12; major type A protein; Mis6; Motch; Motch A; mT14; N1; neurogenic locus notch homolog protein 1; NEXT; NICD; NOTCH; notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch gene homolog 1; Notch homolog 1, translocation-associated; notch receptor 1; Notch1; p300; RP23-306D20.12; RP23-306D20.12-001; Tan1; Translocation-associated notch protein TAN-1; transmembrane receptor Notch1 | |
Mouse | |
Protein A | |
RUO | |
18128, 4851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.