Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Notch1 Monoclonal Antibody (ICN 5B5), Active Motif™

Rat Monoclonal Antibody
$132.55 - $397.65
Specifications
Antigen | Notch1 |
---|---|
Clone | ICN 5B5 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3185
![]() ![]() |
Active Motif
61148 |
10 μg |
Each for $132.55
|
|
|||||
50-199-3184
![]() ![]() |
Active Motif
61147 |
100 μg |
Each for $397.65
|
|
|||||
Description
The notch gene belongs to a family of epidermal growth factor (EGF) like homeotic genes, which encode transmembrane proteins with a variable number of cysteine-rich EGF-like repeats in the extracellular region. Four notch genes have been described in mammals: Notch1, Notch2, Notch3 and Notch4(Int-3), which have been implicated in the differentiation of the nervous system and other structures. The EGF-like proteins Delta and Serrate have been identified as ligands of Notch1. Mature Notch proteins are heterodimeric receptors derived from the cleavage of Notch pre-proteins into an extracellular subunit (NEC) containing multiple EGF-like repeats and a transmembrane subunit including intracellular region (Ntm).Specifications
Notch1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
9930111A19Rik; AOS5; AOVD1; BOS_12152; Drosophila Notch homolog 1 (controlling the the ectodermal and neural cell fate in Drosophila); hN1; lin-12; major type A protein; Mis6; Motch; Motch A; mT14; N1; Neurogenic locus notch homolog protein 1; NEXT; NICD; NOTCH; Notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch gene homolog 1; Notch homolog 1, translocation-associated; NOTCH1; p300; RP23-306D20.12; RP23-306D20.12-001; TAN1; Translocation-associated notch protein TAN-1; transmembrane receptor Notch1 | |
Notch1 | |
IgG2b | |
Protein G | |
Antibody |
ICN 5B5 | |
Western Blot | |
Unconjugated | |
Rat | |
Human, Rat | |
P46531, Q07008 | |
25496, 4851 | |
This Notch1 antibody was raised against a recombinant protein corresponding to the intracellular domain of human Notch1. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Notch1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title