Learn More
Invitrogen™ NOTCH1 Monoclonal Antibody (mN1A)

Description
This antibody is specific for the Notch1 (neurogenic locus notch homolog protein1, Motch A, mT14, p300) protein. This antibody gives a band of ~65 kDa on Western blot analysis using Notch1-IC OP transfected cells. Western blot analysis using Jurkat cells expresses the target band at ~110-120 kDa, with an additional band occasionally observed at ~220kDa. This antibody does not cross-react with Notch 2,3, and 4 as assessed by Western blot analysis.
Specifications
Specifications
| Antigen | NOTCH1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | mN1A |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | Notch1 |
| Gene Accession No. | P46531, Q01705, Q07008 |
| Gene Alias | 9930111A19Rik; AOS5; AOVD1; Drosophila Notch homolog 1 (controlling the the ectodermal and neural cell fate in Drosophila); hN1; lin-12; major type A protein; Mis6; Motch; Motch A; mT14; N1; neurogenic locus notch homolog protein 1; NEXT; NICD; NOTCH; notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch gene homolog 1; Notch homolog 1, translocation-associated; notch receptor 1; Notch1; p300; RP23-306D20.12; RP23-306D20.12-001; Tan1; Translocation-associated notch protein TAN-1; transmembrane receptor Notch1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.