Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch1 Mouse, anti-Human, R718, Clone: MHN1-519, BD OptiBuild™

Catalog No. BDB752187 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752187 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752187 Supplier BD Biosciences Supplier No. 752187
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MHN1-519 monoclonal antibody specifically binds to an extracellular domain of human Notch1. Notch1 is a type 1 transmembrane glycoprotein receptor and member of the Notch family that includes Notch1-Notch4. Notch1 is cleaved in the Golgi and presents as a cell surface heterodimeric receptor. The Notch1 receptor can bind to several membrane-bound ligands including Jagged1, Jagged2, Delta1 and Delta4. Upon ligand binding, Notch1 undergoes proteolytic cleavage that results in the release of the Notch intracellular domain, NICD. NICD translocates to the nucleus where it forms a transcriptional activator complex with various transcriptional factors. These multimeric complexes either positively or negatively regulate the expression of multiple genes including those that orchestrate many facets of embryonic development and the subsequent functioning of multiple organ systems such as the immune, cardiovascular and nervous systems. Within the immune system, Notch signaling significantly affects the development, proliferation, differentiation and survival of numerous cell types including thymocytes and subsets of T and B lymphocytes and dendritic cells. In altered forms, Notch1 has been associated with certain cardiovascular diseases and with some lymphocyte neoplasms.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Notch1
Applications Flow Cytometry
Classification Monoclonal
Clone MHN1-519
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.