Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Notch1a Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP120954
Description
Notch1a Polyclonal specifically detects Notch1a in Zebrafish samples. It is validated for Peptide ELISA.Specifications
Notch1a | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Zebrafish | |
IgG |
ELISA | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:64000 | |
NP_571516.1 | |
NOTCH1 | |
Peptide with sequence C-QNMAPMQHQNISHH corresponding to internal region according to NP_571516.1 . | |
0.1 mg | |
Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
4851 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction