Learn More
Invitrogen™ NOTCH2 Chimeric Recombinant Rabbit Monoclonal Antibody (B9)

Description
Specificity: An engineered ScFv fragment antibody that binds to both mouse mNRR2 and human hNRR2 but not NRR1.
Specifications
Specifications
| Antigen | NOTCH2 Chimeric |
| Applications | ELISA, Inhibition Assays, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | B9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | NOTCH2 |
| Gene Accession No. | O35516, Q04721 |
| Gene Alias | AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.