Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch2 Mouse anti-Human, Brilliant Violet 711, Clone: MHN2-25, BD Optibuild™

Catalog No. BDB742293 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742293 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742293 Supplier BD Biosciences Supplier No. 742293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MHN2-25 monoclonal antibody specifically binds to an extracellular domain of human Notch2. Notch2 is a type 1 transmembrane glycoprotein receptor and member of the Notch family that includes Notch1-Notch4. Notch2 is cleaved in the Golgi and presents as a cell surface heterodimeric receptor. The Notch2 receptor can bind to several membrane-bound ligands including Jagged1, Jagged2, Delta1 and Delta4. Upon ligand binding, Notch2 undergoes proteolytic cleavage that results in the release of the Notch intracellular domain, NICD. NICD translocates to the nucleus where it forms a transcriptional activator complex with various transcriptional factors. These multimeric complexes either positively or negatively regulate the expression of multiple genes including those that orchestrate many facets of embryonic development and the subsequent functioning of multiple organ systems such as the hematopoietic, immune, cardiovascular, hepatic and renal systems. Notch2 is expressed by cells of the lung and brain, as well as cells of the B lineage, thymocytes, and T cells. Abnormalities in Notch2 expression may play a role in the development of B cell lymphomas.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Notch2
Classification Monoclonal
Clone MHN2-25
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias NOTC2, AGS2, hN2, HJCYS, Neurogenic locus notch homolog protein 2
Host Species Mouse
Immunogen Human Notch2 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.