Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOTCH3 Hamster anti-Mouse, Clone: HMN3-133, Invitrogen™
Hamster Monoclonal Antibody
Supplier: Thermo Scientific MA528181
Description
NOTCH3 is the third discovered human homologue of the Drosophila melanogaster type I membrane protein notch. In Drosophila, notch interaction (with its cell-bound ligands delta, serrate) establishes an intercellular signalling pathway that plays a key role in neural development. Homologues of the notch-ligands have also been identified in human. NOTCH3 functions as a receptor for membrane-bound ligands Jagged1, Jagged2, and Delta1 to regulate cell-fate determination. Mutations in NOTCH3 have been identified as the underlying cause of cerebral autosomal domit arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL).
Specifications
NOTCH3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q61982 | |
Hamster | |
Protein G | |
RUO | |
18131 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry | |
HMN3-133 | |
Unconjugated | |
Notch3 | |
Notch3 | |
Mouse Notch 3-Fc fusion protein | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction