Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOTCH3 Mouse anti-Human, Clone:2E4D11, Abnova™

Catalog No. 89334526 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-526 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-526 Supplier Abnova Corporation Supplier No. MAB17841
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human NOTCH3.

This gene encodes the third discovered human homologue of the Drosophilia melanogaster type I membrane protein notch. In Drosophilia, notch interaction with its cell-bound ligands (delta, serrate) establishes an intercellular signalling pathway that plays a key role in neural development. Homologues of the notch-ligands have also been identified in human, but precise interactions between these ligands and the human notch homologues remains to be determined. Mutations in NOTCH3 have been identified as the underlying cause of cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL). [provided by RefSeq]

Specifications

Antigen NOTCH3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E4D11
Conjugate Unconjugated
Description Notch homolog 3 (Drosophila)
Dilution ELISA (1:10000)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene NOTCH3
Gene Alias CADASIL, CASIL
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 47-156 of human NOTCH3 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 4854
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.