Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOTCH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA513203
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513203 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513203 Supplier Invitrogen™ Supplier No. PA513203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with human based on sequence homology.

NOTCH3 is the third discovered human homologue of the Drosophilia melanogaster type I membrane protein notch. In Drosophilia, notch interaction (with its cell-bound ligands delta, serrate) establishes an intercellular signalling pathway that plays a key role in neural development. Homologues of the notch-ligands have also been identified in human. NOTCH3 functions as a receptor for membrane-bound ligands Jagged1, Jagged2, and Delta1 to regulate cell-fate determination. Mutations in NOTCH3 have been identified as the underlying cause of cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOTCH3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Notch3
Gene Accession No. Q61982, Q9UM47
Gene Alias AW229011; CADASIL; CADASIL1; CASIL; hpbk; IMF2; LMNS; N3; Neurogenic locus notch homolog protein 3; notch 3; Notch 3 extracellular truncation; Notch 3 intracellular domain; Notch gene homolog 3; Notch homolog 3; NOTCH3
Gene Symbols Notch3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 2291-2321 amino acids from the C-terminal region of human NOTCH3.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18131, 4854
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.